n-Eicosane ≥99%

Supplier: BeanTown Chemical

Synonyms: N-Eicosane

134330-100G 134330-25G
BT134330-100GEA 68.77 USD
BT134330-100G BT134330-25G
n-Eicosane ≥99%
n-Eicosane

  • Refractive index: 1.443
  • Solid

Formula: H₃C(CH₂)₁₈CH₃
MW: 282.55 g/mol
Boiling Pt: 220 °C/30 mmHg
Melting Pt: 35…37 °C
Density: 0.788 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009344
CAS Number: 112-95-8
EINECS: 204-018-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR