Poly(acrylic acid sodium salt), MW ≈ 5,100

134255-500G 134255-100G
BT134255-500GEA 168.73 USD
BT134255-500G BT134255-100G
Poly(acrylic acid sodium salt), MW ≈ 5,100
CAS: 9003-04-7; MDL No: MFCD00147949; RTECS: WD6826000
Powder; Linear Formula: [-CH2CH(COONa)-]n
Density (g/mL): 0.55
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR