6-Bromoquinoline ≥97%
Supplier: BeanTown Chemical
134135-5G
134135-250MG
134135-1G
BT134135-5GEA
142.28
USD
BT134135-5G
BT134135-250MG
BT134135-1G
6-Bromoquinoline ≥97%
6-Bromoquinoline
- Refractive index: 1.663
- Solid
Formula:
C₉H₆BrN Density: 1.538 g/mL |
MDL Number:
MFCD00024023 CAS Number: 5332-25-2 EINECS: 206-363-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...