Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate

Supplier: BeanTown Chemical

Synonyms: 4-Hydroxy-6-methoxyquinoline-3-carboxylic acid ethyl ester

131730-1G
BT131730-1GEA 505.84 USD
BT131730-1G
Ethyl-4-hydroxy-6-methoxyquinoline-3-carboxylate
Ethyl 4-hydroxy-6-methoxyquinoline-3-carboxylate

  • Solid

Formula: C₁₃H₁₃NO₄
MW: 247.24 g/mol
Melting Pt: 276…277 °C
Density: 1.277 g/mL
MDL Number: MFCD00085617
CAS Number: 77156-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR