Butyl nicotinate ≥98%

Supplier: BeanTown Chemical

Synonyms: Butyl 3-pyridinecarboxylaten-Butyl nicotinateNicotinic acid butyl ester

131220-25G
BT131220-25GEA 98.16 USD
BT131220-25G
Butyl nicotinate ≥98%
Butyl nicotinate

  • RTECS: QT0875000
  • Liquid

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Boiling Pt: 122…123 °C/8 mmHg
Density: 1.06 g/mL
MDL Number: MFCD00006390
CAS Number: 6938-06-3
EINECS: 230-064-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR