4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98%

Supplier: BeanTown Chemical

131185-5G 131185-100G 131185-25G
BT131185-100GEA 399.89 USD
BT131185-100G BT131185-25G BT131185-5G
4,6-Dimethyl-2-methylsulfonylpyrimidine ≥98%
4,6-Dimethyl-2-methylsulfonylpyrimidine

  • Solid

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Melting Pt: 84…86 °C
Density: 1.239 g/mL
MDL Number: MFCD03788286
CAS Number: 35144-22-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR