Niobium nitride

Supplier: BeanTown Chemical

131050-50G 131050-10G
BT131050-50GEA 194.61 USD
BT131050-50G BT131050-10G
Niobium nitride
Niobium nitride

  • Powder

Formula: NNb
Melting Pt: 2573 °C
Density: 8.4 g/mL
MDL Number: MFCD00016239
CAS Number: 24621-21-4
EINECS: 246-362-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR