Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxyethanesulphonic acid sodium saltSodium isethionateSodium hydroxyethyl sulphonate

130570-5KG 130570-1KG
BT130570-5KGEA 331.05 USD
BT130570-5KG BT130570-1KG
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) ≥97%
Isethionic acid sodium salt (Sodium 2-hydroxyethanesulphonate)

  • RTECS: KI7700000
  • Hygroscopic
  • Crystalline

Formula: C₂H₅NaO₄S
MW: 148.11 g/mol
Melting Pt: 191…194 °C
Density: 1.76 g/mL
MDL Number: MFCD00007534
CAS Number: 1562-00-1
EINECS: 216-343-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR