Nitrilotriacetic acid ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: 2,2',2''-Nitrilotriacetic acidNTA

129785-100G 129785-500G
BT129785-100GEA 35.82 USD
BT129785-100G BT129785-500G
Nitrilotriacetic acid ≥98.0% ACS
Nitrilotriacetic acid

  • RTECS: AJ0175000
  • Moisture Sensitive
  • Powder

Formula: C₆H₉NO₆
MW: 191.14 g/mol
Melting Pt: 245 °C (dec.)
Flash Pt: 100 °C (212 °F)
MDL Number: MFCD00004287
CAS Number: 139-13-9
EINECS: 205-355-7
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay 98.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR