N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥95%

Supplier: BeanTown Chemical

Synonyms: Methyl ((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-L-valinate hydrochlorideN-(2'-Cyanobiphenyl-4-ylmethyl) L-valine methyl ester monohydrochloride

129475-25G
BT129475-25GEA 136.05 USD
BT129475-25G
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride ≥95%
N-(2'-Cyanobiphenyl-4-ylmethyl)-L-valinemethylester hydrochloride

  • Solid

Formula: C₂₀H₂₂N₂O₂
MW: 358.87 g/mol
MDL Number: MFCD08460094
CAS Number: 482577-59-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR