Tungsten(V) ethoxide

Supplier: BeanTown Chemical

129325-1G 129325-5G
BT129325-1GEA 44.05 USD
BT129325-1G BT129325-5G
Tungsten(V) ethoxide
Tungsten(V) ethoxide

  • Moisture Sensitive
  • Liquid

Formula: C₁₀H₂₅O₅W
Boiling Pt: 110…115 °C/1 mmHg
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00078041
CAS Number: 26143-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR