Deuterium oxide ≥99.95% (Isotopic)

Synonyms: Heavy waterWater-D2

128765-2EACH 128765-10EACH
BT128765-10EACEA 107.67 USD
BT128765-10EAC BT128765-2EACH
Deuterium oxide ≥99.95% (Isotopic)
Deuterium oxide

  • Refractive index: 1.328
  • Moisture Sensitive
  • Liquid, packaged in ampules, 0.70ml/ampule

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.4 °C
Melting Pt: 3.8 °C
Density: 1.107 g/mL
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR