Nafion® R-1100, sulfonyl fluoride form, resin

Synonyms: Perfluorosulphonyl fluoride-PTFE copolymerPerfluorosulfonyl fluoride-PTFE copolymer

128595-1G 128595-5G
BT128595-1GEA 23.51 USD
BT128595-1G BT128595-5G
Nafion® R-1100, sulfonyl fluoride form, resin
Nafion® R-1100, sulfonyl fluoride form

  • Pellets

Formula: Rf[OCF₂CF(CF₃)₂]nOCF₂CF₂SO₂F
MW: 546.126 g/mol
Melting Pt: >200 °C
MDL Number: MFCD05665455
CAS Number: 26654-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR