5-Acetyluracil ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Acetylpyrimidine-2,4(1H,3H)-dione

128375-25G 128375-5G 128375-1G
BT128375-1GEA 59.71 USD
BT128375-1G BT128375-5G BT128375-25G
5-Acetyluracil ≥98%
5-Acetyluracil

  • Powder

Formula: C₆H₆N₂O₃
MW: 154.11 g/mol
Melting Pt: 278 °C (dec.)
MDL Number: MFCD00151968
CAS Number: 6214-65-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR