3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate
, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane
128140-5G
128140-25G
128140-100G
BT128140-25GEA
49.44
USD
BT128140-25G
BT128140-100G
BT128140-5G
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98%
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate
- Refractive index: 1.419
- Liquid
Formula:
C₁₆H₃₈O₅Si₄ MW: 422.82 g/mol Boiling Pt: 112…115 °C/0.2 mmHg Density: 0.918 g/mL Flash Pt: 201 °C (394 °F) |
MDL Number:
MFCD00053871 CAS Number: 17096-07-0 EINECS: 241-165-0 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...