DL-Ethionine ≥98%

Synonyms: 2-Amino-4-(ethylthio)butanoic acid

127910-1G
BT127910-1GEA 29.79 USD
BT127910-1G
DL-Ethionine ≥98%
DL-Ethionine

  • RTECS: ES6825200
  • Solid

Formula: C₆H₁₃NO₂S
MW: 163.24 g/mol
MDL Number: MFCD00063102
CAS Number: 67-21-0
EINECS: 200-647-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR