1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazole-2-one

127905-1G
BT127905-1GEA 691.76 USD
BT127905-1G
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone ≥97%
1-(1,2,3,6-Tetrahydro-4-pyridyl)-2-benzimidazolinone

  • Solid

Formula: C₁₂H₁₃N₃O
MW: 215.26 g/mol
Melting Pt: 195…200 °C
MDL Number: MFCD00044827
CAS Number: 2147-83-3
EINECS: 218-415-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR