Methyl red sodium salt ACS

Supplier: BeanTown Chemical

Synonyms: Sodium 2-(p-(dimethylamino)phenylazo)benzoate2-(4-Dimethylaminophenylazo)benzoic acid sodium saltMethyl red water soluble

127840-100G 127840-25G
BT127840-100GEA 81.53 USD
BT127840-100G BT127840-25G
Methyl red sodium salt ACS
Methyl red sodium salt

  • Crystalline/Powder

Formula: C₁₅H₁₄N₃NaO₂
MW: 291.28 g/mol
MDL Number: MFCD00002426
CAS Number: 845-10-3
EINECS: 212-682-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR