4-Cyano-4'-pentylbiphenyl ≥98%

Supplier: BeanTown Chemical

Synonyms: 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile4'-Pentyl-4-biphenylcarbonitrile

127685-1G
BT127685-1GEA 85.9 USD
BT127685-1G
4-Cyano-4'-pentylbiphenyl ≥98%
4-Cyano-4'-pentylbiphenyl

  • Refractive index: 1.532
  • Hygroscopic
  • Liquid

Formula: C₁₈H₁₉N
MW: 249.36 g/mol
Boiling Pt: 140…150 °C/0.5 mmHg
Density: 1.008 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00036350
CAS Number: 40817-08-1
EINECS: 255-093-2
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR