1-Pentadecyne ≥98%

127625-1G
BT127625-1GEA 26.21 USD
BT127625-1G
1-Pentadecyne ≥98%
1-Pentadecyne

  • Refractive index: 1.442
  • Liquid

Formula: C₁₅H₂₈
MW: 208.39 g/mol
Density: 0.794 g/mL
MDL Number: MFCD00015081
CAS Number: 765-13-9
EINECS: 212-140-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR