Propiophenone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ethyl phenyl ketone
, Phenyl ethyl ketone
, 1-Phenyl-1-propanone
127155-100G
127155-2.5KG
127155-500G
BT127155-2.5KGEA
182.81
USD
BT127155-2.5KG
BT127155-500G
BT127155-100G
Propiophenone ≥99%
Propiophenone
- RTECS: UG7175000
- Refractive index: 1.526
- Liquid
Formula:
C₆H₅C(O)CH₂CH₃ MW: 134.18 g/mol Boiling Pt: 218 °C Melting Pt: 17…19 °C Density: 1.009 g/mL Flash Pt: 87 °C (188 °F) |
MDL Number:
MFCD00009309 CAS Number: 93-55-0 EINECS: 202-257-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...