Dipraseodymium trioxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

126800-10G 126800-50G
BT126800-10GEA 108.45 USD
BT126800-10G BT126800-50G
Dipraseodymium trioxide ≥99.9% (trace metals basis)
Dipraseodymium trioxide

  • Air Sensitive, Hygroscopic
  • Powder/Lump

Formula: O₃Pr₂
MW: 329.81 g/mol
Melting Pt: 2300 °C
Density: 7.07 g/mL
MDL Number: MFCD00011178
CAS Number: 12036-32-7
EINECS: 234-857-9

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR