N-(Cyclohexylthio)phthalimide ≥98%

Supplier: BeanTown Chemical

Synonyms: Cyclohexyl N-Phthalimidyl Sulfide2-(Cyclohexylthio)isoindoline-1,3-dioneCyclohexyl N-Phthalimidyl Sulphide

144575-25G
BT144575-25GEA 43.32 USD
BT144575-25G
N-(Cyclohexylthio)phthalimide ≥98%
N-(Cyclohexylthio)phthalimide

  • RTECS: TI4290000
  • Solid

Formula: C₁₄H₁₅NO₂S
MW: 261.34 g/mol
Melting Pt: 90 °C
Density: 1.07 g/mL
MDL Number: MFCD00072247
CAS Number: 17796-82-6
EINECS: 241-774-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR