4-Pentynylbenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 5-Phenyl-1-pentyne

130350-25G 130350-5G
BT130350-25GEA 287.58 USD
BT130350-25G BT130350-5G
4-Pentynylbenzene ≥97%
4-Pentynylbenzene

  • RTECS: DA6820000
  • Liquid

Formula: (C₆H₅)CH₂CH₂CH₂CCH
MW: 144.22 g/mol
Boiling Pt: 94 °C/50 mmHg
Density: 0.95 g/mL
Flash Pt: 85 °C (185 °F)
MDL Number: MFCD00039813
CAS Number: 1823-14-9
EINECS: 217-352-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR