2-Carboxyphenylboronic acid ≥95%
Supplier: BeanTown Chemical
Synonyms:
2-Carboxybenzeneboronic acid
, 2-Boronobenzoic acid
126465-250MG
126465-5G
126465-1G
BT126465-1GEA
248.22
USD
BT126465-1G
BT126465-250MG
BT126465-5G
2-Carboxyphenylboronic acid ≥95%
2-Carboxyphenylboronic acid
- Solid
Formula:
C₇H₇BO₄ MW: 165.94 g/mol |
MDL Number:
MFCD01318118 CAS Number: 149105-19-1 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...