N-Acetylneuraminic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: NANASialic acidNeu5AcLactaminic acid(4S,5R,6R,7S,8R)-5-Acetamido-4,6,7,8,9-pentahydroxy-2-oxononanoic acid5-Acetamido-3,5-dideoxy-D-glycero-D-galacto-nonulopyranosonic acid

126220-25MG 126220-100MG
BT126220-25MGEA 34.18 USD
BT126220-25MG BT126220-100MG
N-Acetylneuraminic acid ≥97%
N-Acetylneuraminic acid

  • Powder

Formula: C₁₁H₁₉NO₉
MW: 309.27 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00006620
CAS Number: 131-48-6
EINECS: 205-023-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR