Acetohydroxamic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: N-Hydroxyacetamide

125560-25G 125560-1G 125560-5G
BT125560-1GEA 20.36 USD
BT125560-1G BT125560-25G BT125560-5G
Acetohydroxamic acid ≥98%
Acetohydroxamic acid

  • RTECS: AK8157000
  • Hygroscopic
  • Powder

Formula: C₂H₅NO₂
MW: 75.0666 g/mol
Melting Pt: 88…90 °C
EINECS: 208-913-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR