Azure B (azure I)

Supplier: BeanTown Chemical

Synonyms: Trimethylthionine chlorideC.I. 52010N,N,N′-Trimethylthionin3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chlorideMethylene Azur B

123720-25G 123720-100G 123720-5G
BT123720-5GEA 30.37 USD
BT123720-5G BT123720-100G BT123720-25G
Azure B (azure I)
Azure B (Azure I)

  • Powder

Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Melting Pt: 205…210 °C (dec.)
MDL Number: MFCD00011935
CAS Number: 531-55-5
EINECS: 208-511-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR