Divinylbenzene (mixture of isomers) ≥80% stabilized, Technical Grade

Synonyms: Divinylbenzene

123340-100G 123340-25G 123340-500G
BT123340-25GEA 22.95 USD
BT123340-25G BT123340-100G BT123340-500G
Divinylbenzene (mixture of isomers) ≥80% stabilized, Technical Grade
Divinylbenzene (mixture of isomers)

  • RTECS: CZ9370000
  • Refractive index: 1.561
  • Liquid

Formula: C₁₀H₁₀
MW: 130.19 g/mol
Boiling Pt: 195 °C
Density: 0.914 g/mL
Flash Pt: 64 °C (147 °F)
MDL Number: MFCD01778656
CAS Number: 1321-74-0
EINECS: 215-325-5

Order Now

Specification Test Results

Assay 80%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR