5-Thiazolemethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Thiazol-5-ylmethanolThiazole-5-methanol5-(Hydroxymethyl)thiazole

123245-10G
BT123245-10GEA 49.44 USD
BT123245-10G
5-Thiazolemethanol ≥99%
5-Thiazolemethanol

  • Liquid

Formula: C₄H₅NOS
MW: 115.15 g/mol
Boiling Pt: 249 °C
MDL Number: MFCD04115732
CAS Number: 38585-74-9
EINECS: 414-780-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR