Taurine ≥99%

Supplier: BeanTown Chemical

Synonyms: 2-Aminoethanesulphonic acid2-Aminoethanesulfonic acid

122700-2.5KG 122700-100G 122700-500G
BT122700-500GEA 131.37 USD
BT122700-500G BT122700-2.5KG BT122700-100G
Taurine ≥99%
Taurine

  • RTECS: WX0175000
  • Solid

Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR