18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane18-Crown-6

122390-50G 122390-10G 122390-250G
BT122390-50GEA 176.54 USD
BT122390-50G BT122390-10G BT122390-250G
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) ≥99%
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)

CAS: 17455-13-9; EC No: 241-473-5; MDL No: MFCD00005113; RTECS: MP4500000
Solid; Molecular Formula: C12H24O6; MW: 264.32
Melting Point: 42-45°; Flash point: 110°C (230°F)
Hygroscopic

Formula: C₁₂H₂₄O₆
MW: 264,32 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 37 to 39 °C
Density: 1.237 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR