2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥97%
Supplier: BeanTown Chemical
121725-1G
121725-5G
BT121725-5GEA
415.46
USD
BT121725-5G
BT121725-1G
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥97%
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl
- Crystalline/Solid
Formula:
C₃₀H₄₁F MW: 420.656 g/mol Melting Pt: 129…140 °C |
MDL Number:
MFCD11053361 CAS Number: 102714-93-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...