Ethyl-5-bromonicotinate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 5-bromonicotinate
121170-5G
121170-1G
121170-25G
BT121170-25GEA
762.4
USD
BT121170-25G
BT121170-1G
BT121170-5G
Ethyl-5-bromonicotinate ≥98%
Ethyl 5-bromonicotinate
- Solid
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol Boiling Pt: 86…92 °C/0.5 mmHg Melting Pt: 39…42 °C |
MDL Number:
MFCD00040366 CAS Number: 20986-40-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...