Pifithrin-alpha hydrobromide

Synonyms: 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone HydrobromidePifithrin-alpha

1554-10
10187-524EA 128.22 USD
10187-524
Pifithrin-alpha hydrobromide
Pifithrin-alpha hydrobromide
Formula: C₁₆H₁₈N₂OS·HBr
MW: 286.4·80.9 g/mol
MDL Number: MFCD00417851
CAS Number: 63208-82-2

Order Now

Specification Test Results

CAS 63208-82-2
Purity ≥99%
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR