PluriSIn 1

Supplier: BioVision

Synonyms: NSC 14613Isonicotinic acid 2-PhenylhydrazideN'-Phenylisonicotinohydrazide4-Pyridinecarboxylic acid 2-Phenylhydrazide

AB145629-50MG AB145629-10MG
10190-968EA 245.15 USD
10190-968 10190-970
PluriSIn 1
PluriSIn 1
Formula: C₁₂H₁₁N₃O
MW: 213.24 g/mol
MDL Number: MFCD02344904
CAS Number: 91396-88-2

Order Now

Specification Test Results

CAS 91396-88-2
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR