Forskolin

Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate

1531-5
10187-482EA 119.21 USD
10187-482
Forskolin
Forskolin
Formula: C₂₂H₃₄O₇
MW: 410.51 g/mol
MDL Number: MFCD00082317
CAS Number: 66575-29-9

Order Now

Specification Test Results

CAS 66575-29-9
Purity 98%
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR