TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water

Supplier: BioVision

Synonyms: (p-tert-Octylphenoxy)polyethoxyethanolO-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanolOctoxynolPolyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether

AB286840-100ML
10190-922EA 176.66 USD
10190-922
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
MDL Number: MFCD00132505
CAS Number: 9002-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR