Anisomycin ≥98%

1549-10
10187-510EA 118.17 USD
10187-510
Anisomycin ≥98%
Anisomycin

An activator of p54 and MAP kinases


  • White solid


Synergistic with growth factors and phorbol esters to superinduce cFas and c-Jun, by acting as a potent signaling agonist. Induces apoptosis in the human monoblastoid cell line.


Soluble in DMSO or EtOH


Caution: For research use only, not for use in humans.

Formula: C₁₄H₁₉NO₄
Storage Temperature: Freezer
MDL Number: MFCD00077650
CAS Number: 22862-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR