SB-715992

Synonyms: (R)-N-(3-Aminopropyl)-N-(1-(3-benzyl-7-chloro-4-oxo-3,4-dihydroquinazolin-2-yl)-2-methylpropyl)-4-methylbenzamide

2468-25 2468-5
10190-032EA 180.96 USD
10190-032 10190-034
SB-715992
SB-715992
Formula: C₃₀H₃₃ClN₄O₂
MW: 517,06 g/mol
MDL Number: MFCD22628831
CAS Number: 336113-53-2

Order Now

Specification Test Results

CAS 336113-53-2
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR