ABT-263
Supplier: BioVision
Synonyms:
Navitoclax
, (R)-4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-morpholino-1-(phenylthio)butan-2-yl)amino)-3-((trifluoromethyl)sulfonyl)phenyl)sulfonyl)benzamide
, (R)-4-(4-((4'-Chloro-4,4-dimethyl-3,4,5,6-tetrahydro-[1,1'-biphenyl]-2-yl)methyl)piperazin-1-yl)-N-((4-((4-morpholino-1-(phenylthio)butan-2-yl)amino)-3-((trifluoromethyl)sulphonyl)phenyl)sulphonyl)benzamide
Formula:
C₄₅H₅₅ClF₃N₅O₆S₃ MW: 974.61 g/mol |
MDL Number:
MFCD12756219 CAS Number: 923564-51-6 |
Specification Test Results
CAS | 923564-51-6 |
Purity | ≥98% by HPLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...