ZCL278

Synonyms: 2-(4-Bromo-2-chlorophenoxy)-N-((4-(N-(4,6-dimethylpyrimidin-2-yl)sulfamoyl)phenyl)carbamothioyl)acetamide2-(4-Bromo-2-chlorophenoxy)-N-((4-(N-(4,6-dimethylpyrimidin-2-yl)sulphamoyl)phenyl)carbamothioyl)acetamide

2466-25 2466-5
10190-026EA 531.21 USD
10190-026 10190-024
ZCL278
ZCL278
Formula: C₂₁H₁₉BrClN₅O₄S₂
MW: 584,89 g/mol
MDL Number: MFCD03623090
CAS Number: 587841-73-4

Order Now

Specification Test Results

CAS 587841-73-4
Purity ≥98% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR