WZ4002

Synonyms: N-(3-((5-Chloro-2-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)oxy)phenyl)acrylamide

2461-5 2461-25
10190-010EA 110.91 USD
10190-010 10190-012
WZ4002
WZ4002
Formula: C₂₅H₂₇ClN₆O₃
MW: 494,97 g/mol
MDL Number: MFCD16621244
CAS Number: 1213269-23-8

Order Now

Specification Test Results

CAS 1213269-23-8
Purity ≥98% by HPLC
Form Pale yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR