BAG 956
2456-5
2456-25
10189-992EA
554.56
USD
10189-992
10189-990
BAG 956
BAG 956
CAS Number:
853910-02-8 |
Specification Test Results
CAS | 853910-02-8 |
Purity | ≥98% by HPLC |
Form | Red solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...