BMS-777607

Synonyms: N-(4-((2-Amino-3-chloropyridin-4-yl)oxy)-3-fluorophenyl)-4-ethoxy-1-(4-fluorophenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide

2455-5 2455-25
10191-868EA 554.56 USD
10191-868 10191-866
BMS-777607
BMS-777607
Formula: C₂₅H₁₉ClF₂N₄O₄
MW: 512,89 g/mol
MDL Number: MFCD16495773
CAS Number: 1025720-94-8

Order Now

Specification Test Results

CAS 1025720-94-8
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR