Clioquinol

Synonyms: 5-Chloro-8-hydroxy-7-iodoquinolineClioquinol5-Chloro-7-iodo-8-quinolinol

2454-5G 2454-1G
10189-988EA 75.89 USD
10189-988 10191-864
Clioquinol
Clioquinol
Formula: C₉H₅ClINO
MW: 305.5 g/mol
MDL Number: MFCD00006787
CAS Number: 130-26-7

Order Now

Specification Test Results

CAS 130-26-7
Purity ≥95% by HPLC
Form Tan solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR