3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) ≥98% (by TLC), high purity

Danger

Synonyms: CHAPS

1545-500G 1545-10G 1545-100G 1545-50G 1545-1KG
10187-504EA 3704.37 USD
10187-504 10187-502 10187-500 10187-498 10187-496
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) ≥98% (by TLC), high purity
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

A zwitterionic non-denaturing detergent that protects the native state of proteins, able to disrupt non-specific protein interactions and has less protein aggregation than non-ionic detergents. It is electrically neutral and easily removed by dialysis.

Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081083
CAS Number: 75621-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR