Cycloheximide 100 mM in DMSO

Synonyms: 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,Naramycin AActidione

1041-1
10187-266EA 99.24 USD
10187-266
Cycloheximide 100 mM in DMSO
Cycloheximide
Formula: C₁₅H₂₃NO₄
MW: 281.35 g/mol
MDL Number: MFCD00082346
CAS Number: 66-81-9
https://us.vwr.com/stibo/web/std.lang.all/89/03/2298903.jpg

Order Now

Specification Test Results

CAS 66-81-9
Purity 95%
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR