SNS-314 mesylate

Synonyms: (3-chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea mesyalte1-(3-Chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea methanesulphonate1-(3-Chlorophenyl)-3-(5-(2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl)thiazol-2-yl)urea methanesulfonateN-(3-Chlorophenyl)-N′-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulphonateN-(3-Chlorophenyl)-N′-[5-[2-(thieno[3,2-d]pyrimidin-4-ylamino)ethyl]-2-thiazolyl]urea methanesulfonate

2446-5 2446-25
10191-860EA 554.56 USD
10191-860 10191-858
SNS-314 mesylate
SNS-314 mesylate
Formula: C₁₈H₁₅ClN₆OS₂·CH₄O₃S
MW: 527.04 g/mol
MDL Number: MFCD16621142
CAS Number: 1146618-41-8

Order Now

Specification Test Results

CAS 1057249-41-8
Purity ≥98% by HPLC
Form Light yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR