3-Deazaneplanocin A hydrochloride

Supplier: BioVision

2444-1000 AB145628-250UG
10189-932EA 191.52 USD
10189-932 10191-852
3-Deazaneplanocin A hydrochloride
3-Deazaneplanocin A hydrochloride
Formula: C₁₂H₁₅ClN₄O₃
MW: 298,73 g/mol
MDL Number: MFCD18782644
CAS Number: 120964-45-6

Order Now

Specification Test Results

CAS 120964-45-6
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR